Order Entry
Switzerland
ContactUsLinkComponent
 
Human recombinant Aldolase C (from cells)
  PRSI91-833
 :  
Human recombinant Aldolase C (from cells)
  PRSI91-833
 :  

 

  • Cdt:
    50 µG
  • Type d'enzyme:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Conditions de stockage:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Nom de l'enzyme:
    Aldolase C
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    40.3 kD
  • Sequence:
    Phe2-Tyr364
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 100 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Used for biological assays

 

 

Fructose-bisphosphate aldolase C (ALDOC) belongs to the class I fructose-bisphosphate aldolase family. It is an enzyme that, in humans, is encoded by the ALDOC gene. ALDOC is expressed exclusively in the hippocampus and Purkinje cells of the brain. ALDOC is a glycolytic enzyme which catalyses the reversible aldol cleavage of fructose-1,6-biphosphate and fructose 1-phosphate to dihydroxyacetone phosphate and either glyceraldehyde-3-phosphate or glyceraldehydes respectively

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.