Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant Arginase 1 (from HEK293 cells)
Human recombinant Arginase 1 (from HEK293 cells)
Catalog # PRSI96-034
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant Arginase 1 (from HEK293 cells)
Catalog # PRSI96-034
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Cdt:
    50 µG
  • Type d'enzyme:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Nom de l'enzyme:
    Arginase 1
  • Activité de l'enzyme:
    Measured by the production of urea during the hydrolysis of arginine. The specific activity is >35,000 pmol/min/μg.
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    35.6 kDa
  • Formulation:
    Lyophilized, 50 mM Tris, 150 mM NaCl, pH 7.5
  • Tested applications:
    Western Blot

Specifications

About this item

Arginase-1 (ARG1) is also known as Liver-type arginase, Type I arginase, which belongs to the arginase family. Arginase-1 / ARG1 is a manganese-containing enzyme. The reaction catalysed by this enzyme is: arginine + H2O → ornithine + urea. It is the final enzyme of the urea cycle. Defects in Arginase-1 / ARG1 are the cause of argininemia (ARGIN).