Spécifications
- Nom de l'antigène:Twist Family Bhlh Transcription Factor 1
- Symbole de l'antigène:TWIST1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Formulaire:Liquid
- Synonymes antigène:CRS|CRS1|ACS3|SCS|bHLHa38|TWIST|BPES3|CSO|BPES2
- Concentration:1 mg/mL
- Immunogène:Twist1 antibody was raised against a peptide sequence around aa.18~22 (S-N-S-E-E) derived from Human Twist1.
- Taille:100 µg
- Cdt:100 µG
Spécifications
À propos de cet article
Acts as a transcriptional regulator. Inhibits myogenesis by sequestrating E proteins, inhibiting trans-activation by MEF2, and inhibiting DNA-binding by MYOD1 through physical interaction. This interaction probably involves the basic domains of both proteins. Also represses expression of proinflammatory cytokines such as TNFA and IL1B. Regulates cranial suture patterning and fusion. Activates transcription as a heterodimer with E proteins. Regulates gene expression differentially, depending on dimer composition. Homodimers induce expression of FGFR2 and POSTN while heterodimers repress FGFR2 and POSTN expression and induce THBS1 expression. Heterodimerization is also required for osteoblast differentiation.
Western Blot: 1:500~1:1000
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TWIST1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat