Anti-MAPT Rabbit Polyclonal Antibody
Product Details & Documents
Promotes microtubule assembly and stability, and might be involved in the establishment and maintenance of neuronal polarity. The C-terminus binds axonal microtubules while the N-terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by tau localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The short isoforms allow plasticity of the cytoskeleton whereas the longer isoforms may preferentially play a role in its stabilization.
Western Blot: 1:500~1:1000
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: MAPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Documents
10068-648.pdf
Fiche technique
Spécifications
- Taille:100 µg
- Western blot:Yes
- Cat. No.:PRSI79-540
- Clonalité:Polyclonal
- Formulaire:Liquid
- Immunogène:Tau (Ab-205) antibody was raised against a peptide sequence around aa.203~207 (P-G-T-P-G) derived from Human Tau.
- Nom de l'antigène:Microtubule-Associated Protein tau
- Symbole de l'antigène:MAPT
- Synonymes antigène:PPP1R103|FTDP-17|TAU|PPND|MAPTL|MTBT1|MTBT2|MSTD|DDPAC
- Hôte:Rabbit
- Conjugaison:Unconjugated
- Concentration:1 mg/mL
- Réactivité:Human|Mouse



