Anti-CTDSPL Goat Polyclonal Antibody
PRSI46-827
- Nom de l'antigène:CTD (carboxy-terminal Domain, RNA polymerase II, polypeptide A) small phosphatase-like
- Symbole de l'antigène:CTDSPL
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Goat
- Réactivité:Human
- Formulaire:Liquid
- Synonymes antigène:PSR1|RBSP3|C3orf8|HYA22|SCP3
- Concentration:500 ug/mL
- Immunogène:CTDSPL antibody was raised against a 13 amino acid synthetic peptide near the internal region of CTDSPL.
- Taille:100 µg
- Cdt:100 µG
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 20 kDa band in human placenta lysates after 1 μg/mL antibody staining. Please note that currently we cannot find an explanation in the lite
CTDSPL antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
CTDSPL antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CTDSPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: