Auftragserfassung
Switzerland
ContactUsLinkComponent
 
Anti-CHRNB4 Goat Polyclonal Antibody
Catalog # PRSI46-825
Supplier:  ProSci Inc.
undefined
Anti-CHRNB4 Goat Polyclonal Antibody
Catalog # PRSI46-825
Supplier:  ProSci Inc.

Specifications

  • Symbole de l'antigène:
    CHRNB4
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Goat
  • Réactivité:
    Human
  • Formulaire:
    Liquid
  • Concentration:
    500 ug/mL
  • Immunogène:
    CHRNB4 antibody was raised against a 15 amino acid synthetic peptide near the internal region of CHRNB4.
  • Taille:
    100 µg
  • Cdt:
    100 µG

Specifications

About this item

ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 40 kDa band in human testis lysates after 0.2 μg/mL antibody staining. Please note that currently we cannot find an explanation in the lite

CHRNB4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

CHRNB4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CHRNB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: