Anti-RNF2 Rabbit Polyclonal Antibody
Product Details & Documents
Polycomb group (PcG) of proteins form the multiprotein complexes that are important for the transcription repression of various genes involved in development and cell proliferation. The protein encoded by RNF2 is one of the PcG proteins. It has been shown to interact with, and suppress the activity of, transcription factor CP2 (TFCP2/CP2). This protein was also found to interact with huntingtin interacting protein 2 (HIP2), an ubiquitin-conjugating enzyme, and possess ubiquitin ligase activity.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: RNF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Zebrafish
Documents
10105-868.pdf
Datasheet
Specifications
- Taille:100 µg
- Western blot:Yes
- Formulaire:Lyophilized
- Symbole de l'antigène:RNF2
- Synonymes antigène:HIPI3|RING1B|BAP-1|RING2|DING|BAP1
- Conjugaison:Unconjugated
- Réactivité:Zebrafish|Human|Dog|Rat|Mouse
- Cat. No.:PRSI28-047
- Clonalité:Polyclonal
- Immunogène:A synthetic peptide corresponding a region of human RNF2.
- Nom de l'antigène:Ring Finger Protein 2
- Hôte:Rabbit
- ImmunoChimie:Yes
- ELISA:Yes
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



