Polyester Tags
Fournisseur: Brady
Certificats
À propos de cet article
These two part tags provide enhanced operational control and record keeping. Matching identification numbers are printed on each part.
- Write-on surface accepts marker, pen or pencil
- Weatherproof for either indoor or outdoor use
- Employs ³/₈" rolled brass eyelet to fit over padlock shackles
Polyester 10 mm thickness enhances durability to easily remove chemicals, grease and grime.
Livraison: Nylon cable ties included for easy and secure attachment.
Frequently Bought Together