Drum locks
Drum locks
Steel drum lock set fits 2" and 0.75" NPT bung, 1 kg
N° de catalogue 119-0235
Fournisseur:   JUSTRITE MANUFACTURING
Drum locks
N° de catalogue 119-0235
Fournisseur:  JUSTRITE MANUFACTURING
Drum locks
Steel drum lock set fits 2" and 0.75" NPT bung, 1 kg
Prix
229.00 SFr.
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Drum locks easily install around the drum bung flange over the original bung.
  • Made of corrosion resistant
  • Non-sparking zinc die cast
  • Locks install in seconds to secure contents and reduce theft, pilfering, and sabotage
There is no contact with the drum contents, the potential for content contamination is eliminated and making them suitable for chemical and food processing applications.
Spécifications
  • Désignation:
    Steel drum lock set fits 2" and 0.75" NPT bung, 1 kg