Protective hood, Gridel
DALLCC85278339-AP
- Couleur:fuchsia
- Désignation:Protective hood, made of flame retardant M2
- Cdt:20
Gridel protective hood is designed to isolate the wearer's respiratory tracs when working in surroundings with suspected or low air borne contamination.
- Single use respiratory protection apparatus fed by compressed air
- Comfortable and lightweight
- Can be fitted with a tearable safety strip
- Adjustable air flow
- 360 °C visual field
- Various nipples available
Ideal for use in chemical, nuclear, pharmaceutical, oil and gas industries.
: EN 143, EN 146, EN 270, EN 1073 and EN 1835.