Pipette stand for Gilson pipettes, TRIO™
Catalog # GILSF161405
Supplier: GILSON
Specifications
- Couleur:translucide
- Désignation:Portoir pour pipettes incliné, TRIO™, pour 3 pipettes Gilson (sauf Pipetman Concept®)
- Cdt:1
Specifications
About this item
This handy stand with its angled, transparent design holds up to three single or multi channel Gilson pipettes.
- Made of Temperlite material: Very shock resistant, can be used in chemically aggressive environments
- Protects valuable instruments from shocks and contamination
- Pipettes hang vertically to prevent liquids from running inside the shaft
- Maintain visibility: Its transparent body allows users to see the objects placed behind
- Space saving for a neat, tidy and organized work bench