Bunsen burners, LABOGAZ 206
Catalog # CAMP2000020656
Supplier: CAMPING GAZ
Specifications
- Désignation:Bunsen burner, weight: 380 g
- Cdt:1
Specifications
About this item
For use with Campingaz® pierceable cartridge C 206 GLS. Ideal use in wide range of areas such as laboratories, schools, jewelers, opticians and handicrafts.
- Attached with clip and go installation system which makes changing cartridges easy