PCR mix, ready to use, Taq-&GO™, MP Biomedicals
Catalog # ICNA11EPTAG100
Supplier: MP Biomedicals
CAS Number:
Specifications
- Cdt:1 mL
- Désignation:PCR mix, Taq-&GO™
Specifications
About this item
Taq-&GO™ is a ready to use optimised reaction mix for PCR.
- Significant savings in preparation time
- Reduced pipetting
- Reduced risks of errors and contamination
- Stable at 4 °C
Each mix is provided 5X concentrated and can be used to perform 20 μl or 50 μl reactions. PCR Mastermixes contain all the components for PCR with the exception of template and primers.