ContactUsLinkComponent
Cell culture media, McCoy's 5A, 1X Park and Terasaki's Modification, MP Biomedicals
Cell culture media, McCoy's 5A, 1X Park and Terasaki's Modification, MP Biomedicals
Référence ICNA091692149
Fournisseur:  MP Biomedicals
Numéro CAS:  
Cell culture media, McCoy's 5A, 1X Park and Terasaki's Modification, MP Biomedicals
Référence ICNA091692149
Fournisseur:  MP Biomedicals
Numéro CAS:  
Spécifications
  • Format du milieu:
    Powder
  • Cdt:
    100 mL
  • Désignation:
    McCoy's 5A, 1X Park and Terasaki's Modification
Spécifications
À propos de cet article
The 1X Park and Terasaki's Modification of McCoy's 5A medium is used for the primary culture of cells from a wide range of tissues. Recommended for production of virus in primary cell cultures.
The media is ideally suitable for the propagation of leukocytes, biopsy tissues, a broad range of human and rat normal and transformed cell types, and the most demanding primary and continuous cell lines. This formulation has also be used in the culture of bone marrow, skin, gingiva, kidney, omentum, adrenals, lung, spleen, rat embryos, and other cell types.