Mop cover for handpad holder
Catalog # PPSA2400026
Supplier: PFENNIG REINIGUNGSTECHNIK
Specifications
- Désignation:Special mop laminate
- Longueur:250 mm
- Cdt:1
Specifications
About this item
Special mop laminate made up of PES microfibre for all smooth surfaces. Ideal for removal of persistent decontaminations.