Anti-HTR3E Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-5328R-FITC
Supplier: Bioss
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:5-Hydroxytryptamine (Serotonin) Receptor 3E, Ionotropic
- Symbole de l'antigène:HTR3E
- Clonalité:Polyclonal
- Conjugaison:FITC (Fluorescein Isothiocyanate)
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human
- Adsorption croisée:No
- Formulaire:liquid
- ID de gène:285242
- Synonymes antigène:5-HT3c1|5-HT3E|5-HT3-E
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Température de stockage:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Température de transport:4°C
- Purification:Purified by Protein A
- Cdt:100 µl
Specifications
About this item
HTR3C belongs to the ligand-gated ion channel receptor superfamily. HTR3E is a subunit E of the type 3 receptor for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. Genes encoding subunits C, D and E form a cluster on chromosome 3. An alternative splice variant has been described but its full length sequence has not been determined.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HTR3E
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human