Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:TATA binding Protein TBP
- Symbole de l'antigène:TF2D
- Clonalité:Polyclonal
- Conjugaison:Cy5®
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Adsorption croisée:No
- Formulaire:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Température de stockage:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Température de transport:4°C
- Purification:Purified by Protein A
- Cdt:100 µl
Specifications
About this item
In eukaryotic systems, initiation of transcription from protein-coding genes is a complex process requiring RNA polymerase II and broad families of auxiliary transcription factors. Such factors can be divided into two major functional classes: the basal factors that are required for transcription of all Pol II genes, including TFIIA, TFIIB, TFIID, TFIIE, TFIIF and TFIIH; and sequence-specific factors that regulate gene expression. The basal transcription factors and Pol II form a specific multiprotein complex near the transcription start site by interacting with core promotor elements such as the TATA box generally located 25-30 base pairs upstream of the transcription start site. Binding of TFIID to the TATA element initiates assembly of the other factors into a pre-initiation complex. The TATA-binding subunit of TFIID (designated TFIIDt or TBP) from higher eukaryotes contains a highly conserved 180 amino acid C-terminal domain.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TFIID/TATA binding protein TBP
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat