Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Nerve growth factor
- Symbole de l'antigène:NGF
- Clonalité:Monoclonal
- Clone:BS312
- Conjugaison:Unconjugated
- ELISA:Yes
- Cytométrie en flux:Yes
- Hôte:Mouse
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Réactivité:Human
- Western blot:Yes
- Épitope:A synthetic peptide (C-HTIPQAHWTKLQ, aa: 30-41) of human proNGF protein has been used as the immunogen. The sequence is located on the pro-domain of the proNGF full-length protein.
- Format:Protein G purified IgG.
- Formulaire:Lyophilized
- ID de gène:P01138 NGF_HUMAN
- Synonymes antigène:proNGF|Pro-brain nerve growth factor
- Storage buffer:PBS, pH 7.4, with 3% Trehalose, without preservatives
- Sequence:C-HTIPQAHWTKLQ
- Température de stockage:After reconstitution divide into aliquots and store at −20 °C for a higher stability. Store at 4°C with an appropriate antibacterial agent.
- Température de transport:Room (ambient) temperature.
- Immunogène:A synthetic peptide (C-HTIPQAHWTKLQ, aa: 30-41) of human proNGF protein has been used as the immunogen. The sequence is located on the pro-domain of the proNGF full-length protein.
- Purification:Protein G purified.
- Taille:100 ug
- Cdt:100 µG
Spécifications
À propos de cet article
Nerve growth factor (NGF) is synthesized as a precursor (proNGF) which may be released and have physiological functions to cause cell death. It binds neurotrophin receptor p75 and sortilin and may also be important for the development of nervous system. proNGF is synthesized in target tissues and glia, transported retrogradely and may be released.
Flow Cytometry (2 µg/ 10^6 cells). Immunocytochemistry (1-2 ug/mL), Western Blotting (1-2 ug/mL). Other applications not yet tested. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: NGF
Clonality: Monoclonal
Clone: BS312
Conjugation: Unconjugated
Epitope: A synthetic peptide (C-HTIPQAHWTKLQ, aa: 30-41) of human proNGF protein has been used as the immunogen. The sequence is located on the pro-domain of the proNGF full-length protein.
Host: Mouse
Isotype:
Reactivity: Human