Caméra numérique, AxioCam HRC
N° de catalogue ZEIS4265109901000
Fournisseur: ZEISS
Spécifications
- Modèle:AxioCam HRc
- Résolution:1,4 MP
- Type de sonde:CCD
- Format de sonde:2/3"
- Désignation:Appareil photo numérique
- Seq:999
- Taille de pixel:6,45×6,45 µm
- Interfaces:FireWire IEEE 1394a
- Sonde:Sony ICX 285 Colour CCD
- Cdt:1
Spécifications
À propos de cet article
High resolution camera for complex applications in pathology, developmental biology and material science.
- No rotating fans: Camera works without any vibrations
- Very small dimensions and no external control box
- Large CCD sensor ensures perfect colour accuracy
- High performance at low light intensities
The AxioCam HR can be connected to any Carl Zeiss microscope that has a phototube or TV output.