Trace element solution 2, MP Biomedicals
Référence ICNA091676649
Fournisseur: MP Biomedicals
Numéro CAS:
Spécifications
- Cdt:100 mL
- Désignation:Trace Element Solution 2
Spécifications
À propos de cet article
Trace Element Solution 2 is a 1000X sterile filtered solution, suitable for use with serum-free media; ideal for plant cell culture.
Trace Element Solution 2 is added aseptically to media as a supplement. Prior to use of the media it may be necessary to adjust pH and sterile filter (to reduce contamination).