Foetal bovine serum, South American origin, Corning®
Catalog # MDTC35-079-CV
Supplier: Corning
Specifications
- Désignation:Foetal bovine serum, South American origin, EU compliant
- Cdt:500 mL
Specifications
About this item
Foetal bovine serum sourced in South America and manufactured at facilities approved by the appropriate local authorities.
- Triple sterile filtered (0,1 μm)
- Low endotoxin (<30 EU/ml)
- Low haemoglobin (<30 mg/dl)
Corning® sera products are analysed for quality and performance. Testing includes pH, osmolality, growth promotion, sterility (current USP method), mycoplasma, total protein, endotoxin and haemogloblin levels, and viral contamination. A full biochemical profile is available for each lot upon request.