Reusable half masks, Profile2
Catalog # SCTS5032271
Supplier: SCOTT SAFETY
Specifications
- Type:Profile², helmet harness
- Seq:05
- Taille:M/L
- Environmentally Preferable:
- Cdt:1
Specifications
About this item
Profile² is an advanced twin filter half mask with odourless, lightweight TPE facepiece that combines comfort, protection and low through life costs with exceptionally low breathing resistance. The easy fit bayonet filter mechanism enables fast, secure first time connection.
- Large diameter valves reduce wearer fatigue
- Comfortable head harness and neck buckle provide full support
- Latex and silicone-free to ensure no allergic skin reaction or irritation
- Low profile and swept back filters provide balance and unobstructed field of vision
- Spark, splash and contaminant guards for filters and exhale valve