MICROCHEM® 4000, 406 Chemical Resistant Boot Covers
ANSEGR40W-00406-00
: Ansell
- Pointure:42 - 46
- Applications des tenues:Protection contre les produits chimiques,Pharmaceutical
- Couleur:vert
- Cdt:100 PAIR
These boot covers are made from a multi-layer barrier fabric (external film barrier, non woven inner layer, core high barrier film) that is renowned for its lightweight, yet robust textile feel and exceptional barrier to organic & inorganic chemicals.
- Elastic to top of boot
- Tie fastening
- Ultrasonically welded seams
Typical applications include: Chemical handling, transportation, oil-based mud protection, HAZMAT emergency response, pharmaceutical, mining and agriculture.
: Type 3/4/5, EN 14126, EN 1073-2, EN 1149-5