Anti-POMC Monoclonal Antibody (KLH (Keyhole Limpet Hemocyanin)) [clone: r57]
Catalog # ABNORAB00310
Supplier: Abnova
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Pro-opiomelanocortin
- Symbole de l'antigène:POMC
- Clonalité:Monoclonal
- Clone:r57
- Conjugaison:KLH (Keyhole Limpet Hemocyanin)
- ImmunoChimie:Yes
- Isotype:IgG1 kappa
- Réactivité:Human
- Formulaire:Liquid
- ID de gène:5443
- Storage buffer:In 10 mM PBS.
- Température de stockage:Store at -20° C.
- Immunogène:N-terminal fragment of human POMC conjugated to KLH.
- Purification:Protein A/G purification
- Taille:100 µg
- Cdt:100 µG
Specifications
About this item
Recombinant mouse monoclonal antibody raised against human POMC.
Type: Primary
Antigen: POMC
Clonality: Monoclonal
Clone: r57
Conjugation: KLH (Keyhole Limpet Hemocyanin)
Epitope:
Host:
Isotype: IgG1, kappa
Reactivity: Human