Attaches universelles à quatre pattes
Catalog # RSGA470.000
Supplier: RSG
Specifications
- Largeur de serrage:90 mm
- Cdt:1
Specifications
About this item
Universal clamp made from electrolytically polished stainless steel.
- 4-finger jaws
- Jaws fitted with cork