ContactUsLinkComponent
Accessories for environmental/HazMat racks for small containers
Catalog # CEMG8093
Supplier:  CEMO
undefined
Accessories for environmental/HazMat racks for small containers
Catalog # CEMG8093
Supplier:  CEMO
Specifications
  • Désignation:
    Additional shelf floor, 1000×600×40 mm, 4 kg
  • Cdt:
    1
  • Pour:
    Environmental/HazMat rack 10/20