Eudiometer tubes
Eudiometer tube, complete, includes with reservoir, leveling plate and PVC connecting hose, 1000 ml, 45
N° de catalogue NEUB2591-10-509
Fournisseur: NEUBERT VOLUME GLASSWAERE
Aller à
Product Details & Documents
Eudiometer tubes with spindle tap are suitable for determining the digested behaviour of sludge and biodegradability under anaerobic conditions.
- Made of borosilicate glass 3.3
- With reservoir and leveling plate
- With PVC connecting hose
Certifications: According to DIN 38414 part 8.
Spécifications
- RN:45
- Capacité:1000 ml
- Désignation:Eudiometer tube, complete, includes with reservoir, leveling plate and PVC connecting hose