Light boxes, Slimlite plano LED
Light boxes, Slimlite plano LED
Supplier:   KAISER FOTOTECHNIK
Light boxes, Slimlite plano LED
Supplier:  KAISER FOTOTECHNIK
Certificates
Light boxes, Slimlite plano LED
Light boxes, Slimlite plano LED
2 Options Available Below
Product Details & Documents
Super flat and dimmable daylight light box with build-in rechargeable battery.
  • Uniform and colour precise illumination for viewing, sorting or digitising slides or other transparent originals
  • Various power supply options: Built-in lithium ion battery, the combined mains adapter/charger included or USB cable using a powerbank, computer or car
  • Colour temperature ~5000 Kelvin and colour renditon index of 88 provides for precise colour rendition for inspection and photographic reproduction of originals
  • Scale in cm and inches beside the illuminated area
  • A short touch of the touch switch activates the light source, a longer touch changes brightness continuously
Light source: 23 LEDs, 5000 Kelvin, CRI=88
Built-in Lithium-ion battery (1050 mA) has an operating time of ~6 hours at full power, longer when dimmed.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Light box, illumination area 220×160 mm, 3,6 W, W×D×H 290×200×8 mm, 570 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 180.00
    SFr. 180.00
    1 * 1 ST
       SFr. 180.00
  • Light box, illumination area 320×228 mm, 4,1 W, W×D×H 390×270×8 mm, 1010 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 284.00
    SFr. 284.00
    1 * 1 ST
       SFr. 284.00