Cell detachment solution, Accutase, MP Biomedicals
Catalog # ICNA091000449
Supplier: MP Biomedicals
CAS Number:
Specifications
- Cdt:100 mL
- Désignation:Accutase
Specifications
About this item
Accutase solution is a trypsin replacement that contains both proteolytic and collagenolytic enzymes for cell detachment.
Accutase is effective in detaching primary fibroblasts, endothelial cells, neurons, tumour cell lines, and insect cells. It performs well in detaching cells for analysis of cell surface markers, virus growth assay, and flow cytometry as well as bioreactor scale-up.