ContactUsLinkComponent
Human recombinant alpha-1-Antitrypsin (from E. coli)
 :  Biorbyt
Human recombinant alpha-1-Antitrypsin (from E. coli)
 :  Biorbyt
  • Type de protéine/peptide:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Conditions de stockage:
    SERPINA1 although stable 4 °C for weeks, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA).Please prevent freeze-thaw cycles.
  • Synonyme de protéine:
    AAT|PI|Serpin A1|Rise in alpha 1 antitrypsin occurs as an acute phase response to tissue necrosis and inflammation|Alpha-1 protease inhibitor|Alpha-1-antiproteinase
  • Nom de la protéine/du peptide:
    alpha-1-Antitrypsin
  • Purity:
    Greater than 90% as determined by SDS-PAGE.
  • Sequence:
    MEDPQGDAAQ KTDTSHHDQD HPTFNKITPN LAEFAFSLYR QLAHQSNSTN IFFSPVSIAT AFAMLSLGTK ADTHDEILEG LNFNLTEIPE AQIHEGFQEL LRTLNQPDSQ LQLTTGNGLF LSEGLKLVDK FLEDVKKLYH SEAFTVNFGD TEEAKKQIND YVEKGTQGKI VDLVKELDRD TVFALVNYIF FKGKWERPFE VKDTEEEDFH VDQVTTVKVP MMKRLGMFNI QHCKKLSSWV LLMKYLGNAT AIFFLPDEGK LQHLENELTH DIITKFLENE DRRSASLHLP KLSITGTYDL KSVLGQLGIT KVFSNGADLS GVTEEAPLKL SKAVHKAVLT IDEKGTEAAG AMFLEAIPMS IPPEVKFNKP FVFLMIDQNT KSPLFMGKVV NPTQK.
  • Formulation:
    The protein solution contains 20 mM Tris-HCl pH-7.5, 1mM DTT, 10% glycerol, and 2mM EDTA.
  • Tested applications:
    SDS-PAGE
Frequently Bought Together