Human recombinant MAPK1 (from E. coli)
BIRBORB80800-10
: Biorbyt
:
- Cdt:10 µG
- Type de protéine/peptide:Recombinant
- Source:E. coli
- Species:Human
- Conditions de stockage:Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
- Synonyme de protéine:Mitogen-activated
- Nom de la protéine/du peptide:MAPK1
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Formulation:MAPK1 is supplied containing 50mM Tris-HCL, 150mM NaCl, 2mM DTT, pH 8.0, 100 Units*/mg.
- Tested applications:SDS-PAGE