ContactUsLinkComponent
Human recombinant MIP1A (from E. coli)
Human recombinant MIP1A (from E. coli)
Fournisseur:  Biorbyt
Certificats
Human recombinant MIP1A (from E. coli)
Fournisseur:  Biorbyt
Spécifications
  • Type de protéine/peptide:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Conditions de stockage:
    Store CCL3 at 4 °C if entire vial will be used within 2-4 weeks. Store, frozen at −20 °C for longer periods of time. Please avoid freeze thaw cycles.
  • Synonyme de protéine:
    Small inducible cytokine A3
  • Nom de la protéine/du peptide:
    MIP1A
  • Purity:
    Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Formulation:
    CCL3 His-Tag is supplied in 1x PBS and 50% glycerol.
  • Tested applications:
    SDS-PAGE
Spécifications
Informations associées
Frequently Bought Together