Mouse recombinant RANTES (from E. coli)
Fournisseur: Biorbyt
Certificats
À propos de cet article
Spécifications
- Type de protéine/peptide:Recombinant
- Source:E. coli
- Species:Mouse
- Conditions de stockage:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Synonyme de protéine:Beta chemokine RANTES
- Nom de la protéine/du peptide:RANTES
- Purity:> 97% by SDS-PAGE and HPLC analyses
- Molecular weight:7.9 kDa
- Sequence:SPYGSDTTPC CFAYLSLALP RAHVKEYFYT SSKCSNLAVV FVTRRNRQVCANPEKKWVQE YINYLEMS
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in 3% TFA and methyl cyanides
- Tested applications:SDS-PAGE , HPLC