Mouse recombinant FGF9 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Type de protéine/peptide:Recombinant
- Source:E. coli
- Species:Mouse
- Conditions de stockage:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Synonyme de protéine:Murine Fibroblast Growth Factor-9
- Nom de la protéine/du peptide:FGF9
- Purity:> 95% by SDS-PAGE and HPLC analyses
- Molecular weight:23.3 kDa
- Sequence:MPLGEVGSYF GVQDAVPFGN VPVLPVDSPV LLNDHLGQSE AGGLPRGPAVTDLDHLKGIL RRRQLYCRTG FHLEIFPNGT IQGTRKDHSR FGILEFISIAVGLVSIRGVD SGLYLGMNEK GELYGSEKLT QECVFREQFE ENWYNTYSSNLYKHVDTGRR YYVALNKDGT PREGTRTKRH QKFTHFLPRP VDPDKVPELYKDILSQS
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in 20mM Tris, 500mM NaCl, pH8.5.
- Tested applications:SDS-PAGE , HPLC
Specifications
Frequently Bought Together