Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant HSF2 (from E. coli)
Human recombinant HSF2 (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant HSF2 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Conjugaison:
    Unconjugated
  • Type de protéine/peptide:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Conditions de stockage:
    Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Synonyme de protéine:
    Heat Shock Factor 2
  • Base de données UnitProt:
    Q03933 (Human)
  • Nom de la protéine/du peptide:
    HSF2
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MGSSHHHHHHSSGLVPRGSHMSENKGLETTKNNVVQPVSEEGRKSKSKPDKQLIQYTAFPLLAFL DGNPASSVEQASTTASSEVLSSVDKPIEVDELLDSSLDPEPTQSKLVRLEPLTEAEASEATLFYL CELAPAPLDSDMPLLDS
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 1mM DTT, pH 7.2.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together