Human recombinant ALDOA (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Conjugaison:Unconjugated
- Type de protéine/peptide:Recombinant
- Source:E. coli
- Species:Human
- Conditions de stockage:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Synonyme de protéine:FructoseBisphosphate Aldolase A
- Base de données UnitProt:P04075 (Human)
- Nom de la protéine/du peptide:ALDOA
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:PYQYPALTPEQKKELSDIAHRIVAPGKGILAADESTGSIAKRLQSIGTENTEENRRFYRQLLLTA DDRVNPCIGGVILFHETLYQKADDGRPFPQVIKSKGGVVGIKVDKGVVPLAGTNGETTTQGLDGL SERCAQYKKDGADFAKWRCVLKIGEHTPSALAIMENANVLARYASICQQNGIVPIVEPEILPDGD HDLKRCQYVTEKVLAAVYKALSDHHIYLEGTLLKPNMVTPGHACTQKFSHEEIAMATVTALRRTV PPAVTGITFLSGGQSEEEASINLNAINKCPLLKPWALTFSYGRALQASALKAWGGKKENLKAAQE EYVKRALANSLACQGKYTPSGQAGAAASESLFVSNHAYLEHHHHHH
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 20% Glycerol, pH 8.0.
- Tested applications:SDS-PAGE