ContactUsLinkComponent
Human recombinant CDKN2C (from E. coli)
Human recombinant CDKN2C (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant CDKN2C (from E. coli)
Supplier:  Biorbyt
Specifications
  • Conjugaison:
    Unconjugated
  • Type de protéine/peptide:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Conditions de stockage:
    Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Synonyme de protéine:
    CyclinDependent Kinase 4 Inhibitor C
  • Base de données UnitProt:
    P42773 (Human)
  • Nom de la protéine/du peptide:
    CDKN2C
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MGSSHHHHHHSSGLVPRGSHMAEPWGNELASAAARGDLEQLTSLLQNNVNVNAQNGFGRTALQVM KLGNPEIARRLLLRGANPDLKDRTGFAVIHDAARAGFLDTLQTLLEFQADVNIEDNEGNLPLHLA AKEGHLRVVEFLVKHTASNVGHRNHKGDTACDLARLYGRNEVVSLMQANGAGGATNLQ
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 8.0.
  • Tested applications:
    SDS-PAGE
Specifications
Related Information
Frequently Bought Together