ContactUsLinkComponent
Human recombinant SUMO2 (from E. coli)
 :  Biorbyt
Human recombinant SUMO2 (from E. coli)
 :  Biorbyt
  • Conjugaison:
    Unconjugated
  • Type de protéine/peptide:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Conditions de stockage:
    Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Synonyme de protéine:
    Small UbiquitinRelated Modifier 2
  • Base de données UnitProt:
    P61956 (Human)
  • Nom de la protéine/du peptide:
    SUMO2
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MGSSHHHHHHSSGLVPRGSHMADEKPKEGVKTENNNHINLKVAGQDGSVVQFKIKRHTPLSKLMK AYCERQGLSMRQIRFRFDGQPINETDTPAQLEMEDEDTIDVFQQQTGGVY
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4.
  • Tested applications:
    SDS-PAGE
Frequently Bought Together