ContactUsLinkComponent
Human recombinant LR3IGFI (from E. coli)
 :  Biorbyt
Human recombinant LR3IGFI (from E. coli)
 :  Biorbyt
  • Conjugaison:
    Unconjugated
  • Type de protéine/peptide:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Conditions de stockage:
    Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Activité biologique:
    ED50 is greater than 200 ng/ml as determined by an IGF binding protein assay.
  • Synonyme de protéine:
    InsulinLike Growth Factor I
  • Base de données UnitProt:
    P05019 (Human)
  • Nom de la protéine/du peptide:
    LR3IGFI
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MFPAMPLSSLFVNGPRTLCGAELVDALQFVCGDRGFYFNKPTGYGSSSRRAPQTGIVDECCFRSC DLRRLEMYCAPLKPAKSA
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 300mM HAc-NaAc, pH 6.5.
  • Tested applications:
    SDS-PAGE , FuncS
Frequently Bought Together