Human recombinant CD33 (from HEK293 cells)
Fournisseur: Biorbyt
Certificats
À propos de cet article
Spécifications
- Conjugaison:Unconjugated
- Type de protéine/peptide:Recombinant
- Source:HEK293 cells
- Species:Human
- Conditions de stockage:Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Synonyme de protéine:Myeloid Cell Surface Antigen CD33
- Base de données UnitProt:P20138 (Human)
- Nom de la protéine/du peptide:CD33
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:DPNFWLQVQESVTVQEGLCVLVPCTFFHPIPYYDKNSPVHGYWFREGAIISGDSPVATNKLDQEV QEETQGRFRLLGDPSRNNCSLSIVDARRRDNGSYFFRMERGSTKYSYKSPQLSVHVTDLTHRPKI LIPGTLEPGHSKNLTCSVSWACEQGTPPIFSWLSAAPTSLGPRTTHSSVLIITPRPQDHGTNLTC QVKFAGAGVTTERTIQLNVTYVPQNPTTGIFPGDGSGKQETRAGVVHTSDIEGRMDEPKSCDKTH TCPPCPAPELLGGPSVFLFPPKPKDTLMISRTPEVTCVVVDVSHEDPEVKFNWYVDGVEVHNAKT KPREEQYNSTYRVVSVLTVLHQDWLNGKEYKCKVS NKALPAPIEKTISKAKGQPREPQVYTL PPSREEMTKNQVSLTCLVKGFYPSDIAVEWESNGQPENNYKTTPPVLDSDGSFFLYSKLTVDKSR WQQGNVFSCSVMHEALHNHYTQKSLSLSPGKVDHHHHHH
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 2mM EDTA, pH 7.2.
- Tested applications:SDS-PAGE