ContactUsLinkComponent
Human recombinant CA13 (from E. coli)
 :  Biorbyt
Human recombinant CA13 (from E. coli)
 :  Biorbyt
  • Conjugaison:
    Unconjugated
  • Type de protéine/peptide:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Conditions de stockage:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Synonyme de protéine:
    Carbonic Anhydrase 13
  • Base de données UnitProt:
    Q8N1Q1 (Human)
  • Nom de la protéine/du peptide:
    CA13
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MSRLSWGYREHNGPIHWKEFFPIADGDQQSPIEIKTKEVKYDSSLRPLSIKYDPSSAKIISNSGH SFNVDFDDTENKSVLRGGPLTGSYRLRQVHLHWGSADDHGSEHIVDGVSYAAELHVVHWNSDKYP SFVEAAHEPDGLAVLGVFLQIGEPNSQLQKITDTLDSIKEKGKQTRFTNFDLLSLLPPSWDYWTY PGSLTVPPLLESVTWIVLKQPINISSQQLAKFRSLLCTAEGEAAAFLVSNHRPPQPLKGRKVRAS FHLEHHHHHH
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, pH 7.5.
  • Tested applications:
    SDS-PAGE
Frequently Bought Together