Human recombinant CA1 (from E. coli)
Catalog # BIRBORB49236-1
Supplier: Biorbyt
CAS Number:
Specifications
- Cdt:1 mg
- Conjugaison:Unconjugated
- Type de protéine/peptide:Recombinant
- Source:E. coli
- Species:Human
- Conditions de stockage:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Synonyme de protéine:Carbonic Anhydrase 1
- Base de données UnitProt:P00915 (Human)
- Nom de la protéine/du peptide:CA1
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:ASPDWGYDDKNGPEQWSKLYPIANGNNQSPVDIKTSETKHDTSLKPISVSYNPATAKEIINVGHS FHVNFEDNDNRSVLKGGPFSDSYRLFQFHFHWGSTNEHGSEHTVDGVKYSAELHVAHWNSAKYSS LAEAASKADGLAVIGVLMKVGEANPKLQKVLDALQAIKTKGKRAPFTNFDPSTLLPSSLDFWTYP GSLTHPPLYESVTWIICKESISVSSEQLAQFRSLLSNVEGDNAVPMQHNNRPTQPLKGRTVRASF LEHHHHHH
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, 10% Glycerol, pH 8.0.
- Tested applications:SDS-PAGE