Human recombinant BIN2 (from E. coli)
Marketsource
Fournisseur: Biorbyt
À propos de cet article
3 variantes
Product Details & Documents
Documents
orb49226
Fiche technique
Spécifications de la famille de produits
Spécifications
Spécifications
- Catalog No:
- BIRBORB49226-10
- BIRBORB49226-50
- BIRBORB49226-1
- Nom de la protéine/du peptide:
- BIN2
- BIN2
- BIN2
- Synonyme de protéine:
- Bridging Integrator 2
- Bridging Integrator 2
- Bridging Integrator 2
- Type de protéine/peptide:
- Recombinant
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- Conjugaison:
- Unconjugated
- Unconjugated
- Unconjugated
- Sequence:
- MGSSHHHHHHSSGLVPRGSHMAEGKAGGAAGLFAKQVQKKFSRAQEKVLQKLGKAVETKDERFEQ SASNFYQQQAEGHKLYKDLKNFLSAVKVMHESSKRVSETLQEIYSSEWDGHEELKAIVWNNDLLW EDYEEKLADQAVRTMEIYVAQFSEIKERIAKRGRKLVDYDSARHHLEAVQNAKKKDEAKTAKAEE EFNKAQTVFEDLNQELLEELPILYNSRIGCYVTIFQNISNLRDVFYREMSKLNHNLYEVMSKLEK QHSN
- MGSSHHHHHHSSGLVPRGSHMAEGKAGGAAGLFAKQVQKKFSRAQEKVLQKLGKAVETKDERFEQ SASNFYQQQAEGHKLYKDLKNFLSAVKVMHESSKRVSETLQEIYSSEWDGHEELKAIVWNNDLLW EDYEEKLADQAVRTMEIYVAQFSEIKERIAKRGRKLVDYDSARHHLEAVQNAKKKDEAKTAKAEE EFNKAQTVFEDLNQELLEELPILYNSRIGCYVTIFQNISNLRDVFYREMSKLNHNLYEVMSKLEK QHSN
- MGSSHHHHHHSSGLVPRGSHMAEGKAGGAAGLFAKQVQKKFSRAQEKVLQKLGKAVETKDERFEQ SASNFYQQQAEGHKLYKDLKNFLSAVKVMHESSKRVSETLQEIYSSEWDGHEELKAIVWNNDLLW EDYEEKLADQAVRTMEIYVAQFSEIKERIAKRGRKLVDYDSARHHLEAVQNAKKKDEAKTAKAEE EFNKAQTVFEDLNQELLEELPILYNSRIGCYVTIFQNISNLRDVFYREMSKLNHNLYEVMSKLEK QHSN
- Base de données UnitProt:
- Q9UBW5 (Human)
- Q9UBW5 (Human)
- Q9UBW5 (Human)
- Purity:
- >95% as determined by reducing SDS-PAGE.
- >95% as determined by reducing SDS-PAGE.
- >95% as determined by reducing SDS-PAGE.
- Formulation:
- Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4.
- Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4.
- Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4.
- Conditions de stockage:
- Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Tested applications:
- SDS-PAGE
- SDS-PAGE
- SDS-PAGE
Spécifications
Product Family Options
Informations sur le produit
- CdtDisponibilitéPrix
- Spécifications :Marketsource
Plus
Specifications
Cat. No.BIRBORB49226-10Nom de la protéine/du peptideBIN2Synonyme de protéineBridging Integrator 2Type de protéine/peptideRecombinantSpeciesHumanSourceE. coliConjugaisonUnconjugatedSequenceMGSSHHHHHHSSGLVPRGSHMAEGKAGGAAGLFAKQVQKKFSRAQEKVLQKLGKAVETKDERFEQ SASNFYQQQAEGHKLYKDLKNFLSAVKVMHESSKRVSETLQEIYSSEWDGHEELKAIVWNNDLLW EDYEEKLADQAVRTMEIYVAQFSEIKERIAKRGRKLVDYDSARHHLEAVQNAKKKDEAKTAKAEE EFNKAQTVFEDLNQELLEELPILYNSRIGCYVTIFQNISNLRDVFYREMSKLNHNLYEVMSKLEK QHSNBase de données UnitProtQ9UBW5 (Human)Purity>95% as determined by reducing SDS-PAGE.FormulationLyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4.Conditions de stockageStore at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.Tested applicationsSDS-PAGEMarketsource - Spécifications :Marketsource
Plus
Specifications
Cat. No.BIRBORB49226-50Nom de la protéine/du peptideBIN2Synonyme de protéineBridging Integrator 2Type de protéine/peptideRecombinantSpeciesHumanSourceE. coliConjugaisonUnconjugatedSequenceMGSSHHHHHHSSGLVPRGSHMAEGKAGGAAGLFAKQVQKKFSRAQEKVLQKLGKAVETKDERFEQ SASNFYQQQAEGHKLYKDLKNFLSAVKVMHESSKRVSETLQEIYSSEWDGHEELKAIVWNNDLLW EDYEEKLADQAVRTMEIYVAQFSEIKERIAKRGRKLVDYDSARHHLEAVQNAKKKDEAKTAKAEE EFNKAQTVFEDLNQELLEELPILYNSRIGCYVTIFQNISNLRDVFYREMSKLNHNLYEVMSKLEK QHSNBase de données UnitProtQ9UBW5 (Human)Purity>95% as determined by reducing SDS-PAGE.FormulationLyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4.Conditions de stockageStore at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.Tested applicationsSDS-PAGEMarketsource - Spécifications :Marketsource
Plus
Specifications
Cat. No.BIRBORB49226-1Nom de la protéine/du peptideBIN2Synonyme de protéineBridging Integrator 2Type de protéine/peptideRecombinantSpeciesHumanSourceE. coliConjugaisonUnconjugatedSequenceMGSSHHHHHHSSGLVPRGSHMAEGKAGGAAGLFAKQVQKKFSRAQEKVLQKLGKAVETKDERFEQ SASNFYQQQAEGHKLYKDLKNFLSAVKVMHESSKRVSETLQEIYSSEWDGHEELKAIVWNNDLLW EDYEEKLADQAVRTMEIYVAQFSEIKERIAKRGRKLVDYDSARHHLEAVQNAKKKDEAKTAKAEE EFNKAQTVFEDLNQELLEELPILYNSRIGCYVTIFQNISNLRDVFYREMSKLNHNLYEVMSKLEK QHSNBase de données UnitProtQ9UBW5 (Human)Purity>95% as determined by reducing SDS-PAGE.FormulationLyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4.Conditions de stockageStore at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.Tested applicationsSDS-PAGEMarketsource