Order Entry
Switzerland
ContactUsLinkComponent
 
Recombinant SDF1 alpha (from E. coli)
  BIRBORB82029-10
 :  Biorbyt
 :  
Recombinant SDF1 alpha (from E. coli)
  BIRBORB82029-10
 :  Biorbyt
 :  

 

  • Cdt:
    10 µG
  • Type de protéine/peptide:
    Recombinant
  • Source:
    E. coli
  • Synonyme de protéine:
    C-X-C motif chemokine 12
  • Nom de la protéine/du peptide:
    SDF1 alpha
  • Purity:
    Purified.
  • Formulation:
    Lyophilised.