Portable refractometer for automotive care, Multi-Chek
Référence 635-0509
Fournisseur: REICHERT
Spécifications
- Modèle:Multi-Chek
- Désignation:Digital refractometer for glycol coolants/ battery electrolytes/windshield fluid tester
- Seq:001
- Cdt:1
Spécifications
À propos de cet article
Three-in-one tester for measuring freezing point of engine coolants with ATC (ethylene glycol and propylene glycol); the specific gravity and condition of battery electrolytes and lastly the freezing point of the wind screen washer fluid.