Scoops, non sparking
230 mm, 150 mm, vert
N° de catalogue NEWPTLS290
Fournisseur: NEW PIG
Aller à
Product Details & Documents
Non sparking scoop made from rough, resilient PP that won't rust or corrode. Ideal for handling loose absorbents, powder and contaminated soils.
- Excellent chemical resistance for use in cleaning up loose absorbents
- Non sparking, chemical and corrosion resistant
Spécifications
- Largeur:150 mm
- Couleur:vert
- Longueur:230 mm