Wire basket, rectangular
430×310×220 mm
Catalog # SEMA2659
Supplier: SEMADENI
Jump to:
Product Details & Documents
Made of PP, anthracite.
- Good chemical and temperature resistance
- Space-saving stackable
- With stable, foldable plastic support bracket with locking in vertical position
Specifications
- l×p×h:430×310×220 mm
- Cat. No.:SEMA2659