Stepladder, non-mobile
Non-mobile step ladder, 2, 805×500×600 mm
N° de catalogue PALB2-LAD-NM
Fournisseur: Palbam
Aller à
Product Details & Documents
These step stools are designed for fixed locations where they are not required to be moved often.
- Made up of 304 stainless steel with electropolished finish
Documents
Palbam Class - Catalog 2018
Documentation produit
Spécifications
- No. de marches:2
- Désignation:Non-mobile step ladder
- l×p×h:805×500×600 mm