Anti-Fc CHO cells Antibody [clone: ]
Catalog # ADIPAG35B0002C050
Supplier: AdipoGen Life Sciences
Specifications
- Symbole de l'antigène:Fc
- Hôte:CHO cells
- Réactivité:Human
- Formulaire:Lyophilized.
- Storage buffer:, contains PBS.
- Sequence:Recombinant Fc portion of human IgG1 (aa 105-330).
- Concentration:1 mg/ml (after reconstitution)
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:2 x 50 μg
- Cdt:50 µG
Specifications
About this item
Type:
Antigen: Fc
Clonality:
Clone:
Conjugation:
Epitope:
Host: CHO cells
Isotype:
Reactivity: Human