Anti-SLP-2 Rabbit Polyclonal Antibody [clone: ]
ADIPAG25B0013C050
- Symbole de l'antigène:SLP-2
- Clonalité:Polyclonal
- Hôte:Rabbit
- ImmunoChimie:Yes
- Précipitation immunologique:Yes
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Formulaire:Liquid
- Storage buffer:In PBS containing 0,02% sodium azide.
- Température de stockage:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogène:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taille:100 μg
- Cdt:50 µG
Type:
Antigen: SLP-2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat